Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LRP5
Transcript:
NM_002335.3
Assay Name:
Mutation genome location:
chr.11 68357513-68357770 on build GRCh38
Assay ID
Hs00788087_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
positive regulation of mesenchymal cell proliferation
osteoblast development
glucose catabolic process
endocytosis
cholesterol metabolic process
regulation of blood pressure
positive regulation of cell proliferation
anterior/posterior pattern specification
Wnt signaling pathway
positive regulation of osteoblast proliferation
somatic stem cell population maintenance
extracellular matrix-cell signaling
cell migration involved in gastrulation
cholesterol homeostasis
embryonic digit morphogenesis
regulation of apoptotic process
Wnt signaling pathway involved in dorsal/ventral axis specification
positive regulation of fat cell differentiation
negative regulation of osteoblast differentiation
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
bone remodeling
regulation of bone remodeling
bone marrow development
positive regulation of sequence-specific DNA binding transcription factor activity
anatomical structure regression
retina morphogenesis in camera-type eye
canonical Wnt signaling pathway
bone morphogenesis
branching involved in mammary gland duct morphogenesis
adipose tissue development
cell-cell signaling involved in mammary gland development
regulation of canonical Wnt signaling pathway
regulation of insulin secretion involved in cellular response to glucose stimulus
retinal blood vessel morphogenesis
negative regulation of protein serine/threonine kinase activity
apoptotic process involved in patterning of blood vessels
beta-catenin destruction complex disassembly
protein binding
Wnt-protein binding
toxin transporter activity
Wnt-activated receptor activity
coreceptor activity involved in Wnt signaling pathway
coreceptor activity involved in canonical Wnt signaling pathway
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline