Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
STAT5B
Transcript:
NM_012448.3
Assay Name:
Mutation genome location:
chr.17 42210272-42210531 on build GRCh38
Assay ID
Hs00794409_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
allantoin metabolic process
luteinization
response to hypoxia
natural killer cell differentiation
liver development
citrate metabolic process
2-oxoglutarate metabolic process
succinate metabolic process
oxaloacetate metabolic process
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
isoleucine metabolic process
valine metabolic process
creatine metabolic process
fatty acid metabolic process
acute-phase response
JAK-STAT cascade
female pregnancy
lactation
peptidyl-tyrosine phosphorylation
regulation of steroid metabolic process
taurine metabolic process
lipid storage
regulation of epithelial cell differentiation
response to estradiol
response to lipopolysaccharide
positive regulation of natural killer cell proliferation
positive regulation of natural killer cell differentiation
cellular response to hormone stimulus
T cell differentiation in thymus
prolactin signaling pathway
regulation of multicellular organism growth
positive regulation of multicellular organism growth
positive regulation of activated T cell proliferation
progesterone metabolic process
T cell homeostasis
negative regulation of apoptotic process
positive regulation of interleukin-2 biosynthetic process
response to ethanol
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
negative regulation of erythrocyte differentiation
positive regulation of erythrocyte differentiation
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of natural killer cell mediated cytotoxicity
creatinine metabolic process
development of secondary female sexual characteristics
development of secondary male sexual characteristics
Peyer's patch development
positive regulation of smooth muscle cell proliferation
positive regulation of inflammatory response
positive regulation of cellular component movement
JAK-STAT cascade involved in growth hormone signaling pathway
response to interleukin-2
response to interleukin-4
response to interleukin-15
cellular response to growth factor stimulus
cellular response to epidermal growth factor stimulus
mast cell migration
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein tyrosine kinase activity
signal transducer activity
protein binding
protein phosphatase binding
glucocorticoid receptor binding
protein dimerization activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline