Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PCDHA1
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA10
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA11
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA12
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA13
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA2
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA3
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA4
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA5
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA6
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA7
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA8
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHA9
Transcript:
NM_018904.2
Assay Name:
Mutation genome location:
chr.5 140884173-140884422 on build GRCh38
Assay transcription exon location:
Within Exon 1
Assay ID
Hs00813147_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
nervous system development
calcium ion binding
protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline