Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATP1A2
Transcript:
NM_000702.3
Assay Name:
Mutation genome location:
chr.1 160120865-160121118 on build GRCh38
Assay ID
Hs00829716_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter uptake
regulation of the force of heart contraction
regulation of respiratory gaseous exchange by neurological system process
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
regulation of smooth muscle contraction
regulation of striated muscle contraction
regulation of blood pressure
adult locomotory behavior
visual learning
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
ATP hydrolysis coupled proton transport
regulation of vasoconstriction
cellular potassium ion homeostasis
ion transmembrane transport
response to nicotine
sodium ion export from cell
locomotion
negative regulation of heart contraction
negative regulation of striated muscle contraction
ATP metabolic process
negative regulation of cytosolic calcium ion concentration
regulation of glutamate uptake involved in transmission of nerve impulse
regulation of synaptic transmission, glutamatergic
relaxation of cardiac muscle
cardiac muscle contraction
cellular response to mechanical stimulus
cellular response to steroid hormone stimulus
regulation of cardiac muscle cell contraction
membrane repolarization
membrane depolarization during cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
negative regulation of calcium ion transmembrane transport
negative regulation of calcium:sodium antiporter activity
response to glycoside
regulation of cardiac conduction
potassium ion import across plasma membrane
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
drug binding
ATPase activity
potassium ion binding
sodium ion binding
chaperone binding
steroid hormone binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline