Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175101.3 | NP_780310.1 | 2 | 349 | ||
GenBank mRNA | AK020058.1 | - | 2 | 320 | ||
AK075595.1 | - | 2 | 350 | |||
AK167819.1 | - | 2 | 198 | |||
AK206599.1 | - | 2 | 95 | |||
AK213319.1 | - | 2 | 95 | |||
AK218418.1 | - | 2 | 95 | |||
BC004641.1 | - | 2 | 345 | 3157147 |