Search
Search

ICAM4| Gene Symbol: ICAM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039132.2 | NP_001034221.1 | 3 | 830 | ||
| NM_001544.4 | NP_001535.1 | 3 | 907 | |||
| NM_022377.3 | NP_071772.1 | 2 | 1054 | |||
| GenBank mRNA | BC000046.2 | - | 3 | 823 | 3505509 | |
| BC029364.1 | - | 3 | 907 | 4659161 | ||
| BT009816.1 | - | 3 | 784 | |||
| L27670.1 | - | 2 | 1014 | |||
| L27671.1 | - | 3 | 867 | |||
| S78853.1 | - | 1 | 69 | |||