Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 8 | 1003 | ||
NM_002718.4 | NP_002709.2 | 9 | 3438 | |||
NM_181897.2 | NP_871626.1 | 8 | 1293 | |||
XM_006713686.3 | XP_006713749.1 | 9 | 3849 | |||
XM_011512956.2 | XP_011511258.1 | 9 | 3613 | |||
XM_017006787.1 | XP_016862276.1 | 8 | 3171 | |||
GenBank mRNA | AK293012.1 | - | 9 | 3418 | ||
AK298059.1 | - | 8 | 932 | |||
AK299544.1 | - | 8 | 963 | |||
AK314243.1 | - | 9 | 3366 | |||
AK316258.1 | - | 8 | 1246 | |||
AL389975.1 | - | 6 | 521 | |||
BC065531.1 | - | 9 | 3385 | 6161260 | ||
L07590.1 | - | 9 | 3325 | |||
L12146.1 | - | 8 | 1218 |