Search
Search

BSPRY| Gene Symbol: BSPRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317943.1 | NP_001304872.1 | 6 | 1325 | ||
| NM_001317944.1 | NP_001304873.1 | 5 | 1185 | |||
| NM_017688.2 | NP_060158.2 | 6 | 1310 | |||
| XM_006717149.3 | XP_006717212.1 | 6 | 1298 | |||
| GenBank mRNA | AJ276691.1 | - | 6 | 1291 | ||
| AK000157.1 | - | 6 | 1036 | |||
| AK092607.1 | - | 6 | 1301 | |||
| BC001477.2 | - | 5 | 1194 | |||