Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024881.4 | NP_079157.3 | 6 | 2041 | ||
GenBank mRNA | AK027699.1 | - | 6 | 1835 | ||
AK027850.1 | - | 4 | 1452 | |||
AK075355.1 | - | 4 | 1480 | |||
BC014557.2 | - | 3 | 1388 | |||
BC062562.1 | - | 6 | 2041 | 6598965 | ||
BX640756.1 | - | 6 | 1652 |