Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GOSR2 SIRNA Assays ›
          Gene Symbol:
          GOSR2
          Gene Name:
          golgi SNAP receptor complex member 2
          Gene Aliases:
          Bos1, EPM6, GS27
          Chromosome Location:
          Chr.17: 46923086 - 46975519 on Build GRCh38
          UniGene ID:
          Hs.463278
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112311

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GOSR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012511.2 NP_001012529.1 4, 5 407
          NM_001321133.1 NP_001308062.1 4, 5 407
          NM_004287.4 NP_004278.2 4, 5 407
          NM_054022.3 NP_473363.1 4, 5 407
          XM_006722190.3 XP_006722253.1 4, 5 416
          XM_011525501.2 XP_011523803.1 4, 5 416
          XM_011525502.2 XP_011523804.1 4, 5 416
          XM_017025378.1 XP_016880867.1 4, 5 413
          XM_017025379.1 XP_016880868.1 4, 5 407
          XM_017025380.1 XP_016880869.1 4, 5 764
          XM_017025381.1 XP_016880870.1 4, 5 390
          XM_017025382.1 XP_016880871.1 4, 5 742
          XM_017025383.1 XP_016880872.1 4, 5 416
          XM_017025384.1 XP_016880873.1 4, 5 438
          XM_017025385.1 XP_016880874.1 4, 5 413
          XM_017025388.1 XP_016880877.1 4, 5 416
          XM_017025389.1 XP_016880878.1 4, 5 423
          XM_017025390.1 XP_016880879.1 4, 5 768
          XM_017025391.1 XP_016880880.1 4, 5 766
          GenBank mRNA AF229796.1 - 4, 5 362
          AK290890.1 - 4, 5 382
          BC009710.2 - 4, 5 378 3890308
          BC034762.1 - 4, 5 350 5185659
          BG718145.1 - 4, 5 401

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to vacuole
          ER to Golgi vesicle-mediated transport
          intra-Golgi vesicle-mediated transport
          Golgi to vacuole transport
          IRE1-mediated unfolded protein response
          retrograde transport, endosome to Golgi
          COPII vesicle coating
          vesicle fusion with Golgi apparatus
          membrane fusion
          SNARE binding
          transporter activity
          SNAP receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0