Search
Search

GABRA3| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000808.3 | NP_000799.1 | 10 | 1851 | ||
| XM_006724811.2 | XP_006724874.1 | 9 | 1639 | |||
| GenBank mRNA | BC028629.1 | - | 10 | 1850 | 5261326 | |
| KJ620007.1 | - | 16 | 2480 | |||