Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARNT SIRNA Assays ›
Gene Symbol:
ARNT
Gene Name:
aryl hydrocarbon receptor nuclear translocator
Gene Aliases:
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
Chromosome Location:
Chr.1: 150809705 - 150876768 on Build GRCh38
UniGene ID:
Hs.632446
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114177
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARNT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197325.1
NP_001184254.1
4
416
NM_001286035.1
NP_001272964.1
5
472
NM_001286036.1
NP_001272965.1
4
416
NM_001668.3
NP_001659.1
4
416
NM_178427.2
NP_848514.1
4
416
XM_005245151.1
XP_005245208.1
4
358
XM_005245153.1
XP_005245210.1
4
358
XM_005245154.3
XP_005245211.1
5
386
XM_005245157.1
XP_005245214.1
4
358
XM_011509542.2
XP_011507844.1
4
419
XM_011509543.2
XP_011507845.1
5
414
XM_011509544.2
XP_011507846.1
4
383
XM_011509545.2
XP_011507847.1
4
419
XM_011509546.1
XP_011507848.1
3
134
XM_011509547.2
XP_011507849.1
4
419
XM_017001288.1
XP_016856777.1
5
261
XM_017001289.1
XP_016856778.1
5
261
XM_017001290.1
XP_016856779.1
5
260
XM_017001291.1
XP_016856780.1
5
262
XM_017001292.1
XP_016856781.1
5
262
XM_017001293.1
XP_016856782.1
5
264
XM_017001294.1
XP_016856783.1
4
358
XM_017001295.1
XP_016856784.1
4
358
XM_017001296.1
XP_016856785.1
4
367
GenBank mRNA
AB209877.1
-
4
227
AF001307.1
-
4
270
AK223459.1
-
280
AK290177.1
-
4
355
AK291705.1
-
4
258
AK293027.1
-
4
416
AK304772.1
-
4
250
AK313780.1
-
4
357
AL834279.1
-
5
411
BC041121.1
-
4
230
BC060838.1
-
4
351
5300334
M69238.1
-
4
271
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
embryonic placenta development
positive regulation of endothelial cell proliferation
transcription, DNA-templated
positive regulation of vascular endothelial growth factor production
cell differentiation
intracellular receptor signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of protein sumoylation
mRNA transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
positive regulation of erythrocyte differentiation
positive regulation of glycolytic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hormone biosynthetic process
regulation of transcription from RNA polymerase II promoter in response to hypoxia
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
aryl hydrocarbon receptor activity
protein binding
transcription factor binding
aryl hydrocarbon receptor binding
enhancer binding
sequence-specific DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0