Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123662
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
22
3541
NM_032291.3
NP_115667.2
25
4318
XM_005271264.3
XP_005271321.1
25
4152
XM_005271268.3
XP_005271325.1
23
3567
XM_005271270.4
XP_005271327.1
22
3874
XM_006710961.2
XP_006711024.1
25
4158
XM_006710966.2
XP_006711029.1
24
4092
XM_006710967.2
XP_006711030.1
24
4080
XM_006710969.2
XP_006711032.1
24
4068
XM_006710971.2
XP_006711034.1
24
3657
XM_006710972.2
XP_006711035.1
24
3651
XM_006710973.2
XP_006711036.1
22
3633
XM_006710974.2
XP_006711037.1
21
3549
XM_011542291.1
XP_011540593.1
24
3814
XM_011542292.1
XP_011540594.1
23
3754
XM_011542293.1
XP_011540595.1
20
3483
XM_017002505.1
XP_016857994.1
26
4206
XM_017002506.1
XP_016857995.1
27
4329
XM_017002507.1
XP_016857996.1
26
4381
XM_017002508.1
XP_016857997.1
27
4327
XM_017002509.1
XP_016857998.1
24
4182
XM_017002510.1
XP_016857999.1
25
4305
XM_017002511.1
XP_016858000.1
26
4263
XM_017002512.1
XP_016858001.1
26
4136
XM_017002513.1
XP_016858002.1
25
4315
XM_017002514.1
XP_016858003.1
25
4134
XM_017002515.1
XP_016858004.1
23
4110
XM_017002516.1
XP_016858005.1
24
4233
XM_017002517.1
XP_016858006.1
25
4202
XM_017002518.1
XP_016858007.1
25
4190
XM_017002519.1
XP_016858008.1
25
4215
XM_017002520.1
XP_016858009.1
23
4056
XM_017002521.1
XP_016858010.1
23
4008
XM_017002522.1
XP_016858011.1
24
4130
XM_017002523.1
XP_016858012.1
24
4154
XM_017002524.1
XP_016858013.1
24
3797
XM_017002525.1
XP_016858014.1
23
3736
XM_017002526.1
XP_016858015.1
22
3615
XM_017002527.1
XP_016858016.1
23
3639
XM_017002528.1
XP_016858017.1
23
3725
XM_017002529.1
XP_016858018.1
23
3591
XM_017002530.1
XP_016858019.1
22
3567
XM_017002531.1
XP_016858020.1
21
3567
XM_017002532.1
XP_016858021.1
22
3561
XM_017002533.1
XP_016858022.1
22
3669
XM_017002534.1
XP_016858023.1
21
3543
XM_017002535.1
XP_016858024.1
22
3657
XM_017002536.1
XP_016858025.1
22
3507
XM_017002537.1
XP_016858026.1
21
3596
GenBank mRNA
AB210039.1
-
25
4101
AK125044.1
-
8
3085
AL136561.1
-
25
3992
BX640813.1
-
22
3518
CR749541.1
-
22
3729
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0