Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BUD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032725.3 | NP_116114.1 | 4 | 958 | ||
XM_011543035.2 | XP_011541337.1 | 4 | 1532 | |||
GenBank mRNA | AK289635.1 | - | 4 | 947 | ||
AK291111.1 | - | 4 | 947 | |||
AK311068.1 | - | 4 | 947 | |||
BC006350.2 | - | 4 | 948 | 4111572 |