Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFP36 SIRNA Assays ›
Gene Symbol:
ZFP36
Gene Name:
ZFP36 ring finger protein
Gene Aliases:
G0S24, GOS24, NUP475, RNF162A, TIS11, TTP, zfp-36
Chromosome Location:
Chr.19: 39406847 - 39409412 on Build GRCh38
UniGene ID:
Hs.534052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13032
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFP36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003407.3
NP_003398.2
2
1306
GenBank mRNA
BC009693.1
-
2
1288
3900312
M63625.1
-
2
1257
M92843.1
-
2
1307
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
nuclear-transcribed mRNA poly(A) tail shortening
mRNA catabolic process
response to wounding
regulation of keratinocyte proliferation
nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
regulation of tumor necrosis factor production
miRNA mediated inhibition of translation
p38MAPK cascade
response to starvation
regulation of mRNA stability
cellular response to fibroblast growth factor stimulus
positive regulation of fat cell differentiation
regulation of keratinocyte differentiation
negative regulation of erythrocyte differentiation
mRNA transport
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
3'-UTR-mediated mRNA destabilization
3'-UTR-mediated mRNA stabilization
cellular response to lipopolysaccharide
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to glucocorticoid stimulus
cellular response to granulocyte macrophage colony-stimulating factor stimulus
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
regulation of keratinocyte apoptotic process
positive regulation of intracellular mRNA localization
DNA binding
single-stranded RNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
AU-rich element binding
enzyme binding
protein kinase binding
C-C chemokine binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0