Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCHL5 SIRNA Assays ›
          Gene Symbol:
          UCHL5
          Gene Name:
          ubiquitin C-terminal hydrolase L5
          Gene Aliases:
          CGI-70, INO80R, UCH-L5, UCH37
          Chromosome Location:
          Chr.1: 193012366 - 193060140 on Build GRCh38
          UniGene ID:
          Hs.145469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 134742

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UCHL5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199261.1 NP_001186190.1 5 533
          NM_001199262.1 NP_001186191.1 5 533
          NM_001199263.1 NP_001186192.1 5 533
          NM_015984.3 NP_057068.1 5 533
          NR_037607.1 5 799
          XM_005245244.4 XP_005245301.1 5 616
          XM_005245246.4 XP_005245303.1 5 616
          XM_005245247.4 XP_005245304.1 5 468
          XM_005245248.4 XP_005245305.1 5 491
          XM_005245249.4 XP_005245306.1 5 387
          XM_006711366.3 XP_006711429.1 5 616
          XM_006711367.3 XP_006711430.1 5 616
          XM_006711368.3 XP_006711431.1 5 616
          XM_006711369.2 XP_006711432.1 5 616
          XM_006711370.3 XP_006711433.3 5 569
          XM_006711371.3 XP_006711434.1 5 387
          XM_011509604.1 XP_011507906.1 5 616
          XM_011509607.2 XP_011507909.1 5 427
          XM_011509608.2 XP_011507910.1 6 788
          XM_011509609.2 XP_011507911.1 6 692
          XM_017001429.1 XP_016856918.1 5 616
          XM_017001430.1 XP_016856919.1 5 616
          XM_017001431.1 XP_016856920.1 5 616
          XM_017001432.1 XP_016856921.1 5 616
          XM_017001433.1 XP_016856922.1 5 616
          XM_017001434.1 XP_016856923.1 5 569
          XM_017001435.1 XP_016856924.1 5 569
          XM_017001436.1 XP_016856925.1 6 788
          XM_017001437.1 XP_016856926.1 6 788
          XM_017001438.1 XP_016856927.1 5 411
          XM_017001439.1 XP_016856928.1 5 832
          XM_017001440.1 XP_016856929.1 5 796
          XM_017001441.1 XP_016856930.1 5 772
          XM_017001442.1 XP_016856931.1 5 387
          XM_017001443.1 XP_016856932.1 5 387
          XM_017001444.1 XP_016856933.1 5 781
          GenBank mRNA AF147717.1 - 5 531
          AF151828.1 - 5 413
          AF157320.1 - 5 548
          AK024751.1 - 5 479
          AK225794.1 - 5 466
          AK301381.1 - 5 799
          AK314445.1 - 5 477
          AK316064.1 - 4 371
          BC015521.1 - 5 466 3912584
          BC025369.2 - 5 511 3937407
          BT006790.1 - 5 400
          DA255130.1 - 5 533

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          negative regulation of endopeptidase activity
          protein deubiquitination
          lateral ventricle development
          midbrain development
          forebrain morphogenesis
          regulation of proteasomal protein catabolic process
          thiol-dependent ubiquitin-specific protease activity
          endopeptidase inhibitor activity
          protein binding
          poly(A) RNA binding
          proteasome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0