Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IKZF1 SIRNA Assays ›
          Gene Symbol:
          IKZF1
          Gene Name:
          IKAROS family zinc finger 1
          Gene Aliases:
          CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
          Chromosome Location:
          Chr.7: 50303453 - 50405101 on Build GRCh38
          UniGene ID:
          Hs.435949
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135570

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IKZF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001220765.2 NP_001207694.1 7 1530
          NM_001220767.2 NP_001207696.1 6 1111
          NM_001220768.2 NP_001207697.1 5 1141
          NM_001220770.2 NP_001207699.1 5 985
          NM_001220771.2 NP_001207700.1 4 973
          NM_001291837.1 NP_001278766.1 7 1503
          NM_001291838.1 NP_001278767.1 7 1368
          NM_001291839.1 NP_001278768.1 6 1047
          NM_001291840.1 NP_001278769.1 3 712
          NM_001291841.1 NP_001278770.1 5 973
          NM_001291842.1 NP_001278771.1 5 943
          NM_001291843.1 NP_001278772.1 4 847
          NM_001291844.1 NP_001278773.1 4 817
          NM_006060.5 NP_006051.1 8 1656
          XM_011515058.2 XP_011513360.1 9 1546
          XM_011515059.2 XP_011513361.1 9 1518
          XM_011515060.2 XP_011513362.1 9 4696
          XM_011515061.2 XP_011513363.1 8 1484
          XM_011515062.2 XP_011513364.1 9 1826
          XM_011515063.2 XP_011513365.1 9 1743
          XM_011515064.2 XP_011513366.1 9 5374
          XM_011515065.2 XP_011513367.1 9 1511
          XM_011515066.2 XP_011513368.1 9 1615
          XM_011515067.2 XP_011513369.1 8 1422
          XM_011515068.2 XP_011513370.1 8 1683
          XM_011515069.2 XP_011513371.1 8 5311
          XM_011515070.2 XP_011513372.1 7 1361
          XM_011515071.2 XP_011513373.1 8 1285
          XM_011515072.2 XP_011513374.1 7 1223
          XM_011515073.2 XP_011513375.1 8 1565
          XM_011515074.2 XP_011513376.1 7 1196
          XM_011515075.2 XP_011513377.1 7 1505
          XM_011515076.2 XP_011513378.1 7 1449
          XM_011515077.2 XP_011513379.1 6 1100
          XM_011515078.2 XP_011513380.1 6 1073
          XM_017011667.1 XP_016867156.1 8 1482
          XM_017011668.1 XP_016867157.1 7 1189
          GenBank mRNA AF432219.1 - 2 651
          AK303586.1 - 7 1327
          AK308411.1 - 7 1460
          AY377974.1 - 7 1428
          BC018349.1 - 7 1454 4338834
          BT009836.1 - 7 1262
          JX459579.1 - 6 803
          S80876.1 - 6 1616
          U40462.1 - 8 1556

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107237, XM_006251495, XM_008770318, XM_008770312, XM_006251496, and others.
          This siRNA also targets the following transcripts in Mouse: L03547, AK036833, AK142856, AY536269, AK077815, and others.

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          cell cycle
          mesoderm development
          covalent chromatin modification
          lymphocyte differentiation
          erythrocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription regulatory region DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0