Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005814.2 | NP_005805.1 | 7 | 1742 | ||
XM_017000005.1 | XP_016855494.1 | 8 | 1570 | |||
GenBank mRNA | BC069761.1 | - | 7 | 1742 | 7262565 | |
BC069789.1 | - | 7 | 1742 | 7262482 | ||
BC107164.1 | - | 7 | 1697 | 40034324 | ||
BC107165.1 | - | 7 | 1699 | 40034331 | ||
U79725.1 | - | 7 | 1742 |