Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICK1 SIRNA Assays ›
          Gene Symbol:
          PICK1
          Gene Name:
          protein interacting with PRKCA 1
          Gene Aliases:
          PICK, PRKCABP
          Chromosome Location:
          Chr.22: 38056311 - 38075704 on Build GRCh38
          UniGene ID:
          Hs.180871
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138240

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PICK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039583.1 NP_001034672.1 8 816
          NM_001039584.1 NP_001034673.1 8 763
          NM_012407.3 NP_036539.1 8 914
          XM_006724379.2 XP_006724442.1 7 700
          XM_006724380.2 XP_006724443.1 7 619
          XM_011530517.2 XP_011528819.1 7 733
          XM_017029090.1 XP_016884579.1 8 878
          XM_017029091.1 XP_016884580.1 8 929
          XM_017029092.1 XP_016884581.1 8 841
          XM_017029093.1 XP_016884582.1 6 586
          XM_017029094.1 XP_016884583.1 7 892
          XM_017029095.1 XP_016884584.1 10 1505
          GenBank mRNA AB026491.1 - 8 727
          AF231710.1 - 7 536
          AK000875.1 - 8 670
          AK092818.1 - 8 764
          AK222640.1 - 8 762
          AK299684.1 - 8 611
          AK308792.1 - 7 521
          AK310629.1 - 9 1012
          AL049654.1 - 8 751
          BC017561.2 - 8 732 4026028
          CR456550.1 - 8 637

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          protein phosphorylation
          retrograde vesicle-mediated transport, Golgi to ER
          protein kinase C-activating G-protein coupled receptor signaling pathway
          monoamine transport
          glial cell development
          regulation of Arp2/3 complex-mediated actin nucleation
          negative regulation of Arp2/3 complex-mediated actin nucleation
          cellular response to decreased oxygen levels
          cellular response to glucose starvation
          DNA methylation involved in embryo development
          DNA methylation involved in gamete generation
          receptor clustering
          neuronal ion channel clustering
          long term synaptic depression
          dendritic spine organization
          dendritic spine maintenance
          G-protein coupled receptor binding
          protein kinase C binding
          receptor binding
          protein binding
          protein C-terminus binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          identical protein binding
          metal ion binding
          actin filament binding
          Arp2/3 complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bnpf6:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0