Search
Search

NSMAF| Gene Symbol: NSMAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144772.1 | NP_001138244.1 | 3 | 361 | ||
| NM_003580.3 | NP_003571.2 | 3 | 421 | |||
| GenBank mRNA | AK292679.1 | - | 3 | 301 | ||
| AK294009.1 | - | 3 | 361 | |||
| AK313991.1 | - | 3 | 332 | |||
| BC041124.1 | - | 3 | 403 | 5785460 | ||
| DR005814.1 | - | 3 | 427 | |||
| X96586.1 | - | 3 | 219 | |||