Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006418.4 | NP_006409.3 | 3 | 542 | ||
GenBank mRNA | AK000683.1 | - | 4 | 441 | ||
AK300947.1 | - | 3 | 422 | |||
AY358567.1 | - | 3 | 459 | |||
BC047740.1 | - | 2 | 173 | |||
BC117329.1 | - | 3 | 524 | 40125880 | ||
DA928098.1 | - | 3 | 547 |