Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAOK3 SIRNA Assays ›
          Gene Symbol:
          TAOK3
          Gene Name:
          TAO kinase 3
          Gene Aliases:
          DPK, JIK, MAP3K18
          Chromosome Location:
          Chr.12: 118149801 - 118372950 on Build GRCh38
          UniGene ID:
          Hs.644420
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 140330

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAOK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016281.3 NP_057365.3 11, 12 1317
          XM_005253897.1 XP_005253954.1 11, 12 1288
          XM_005253898.1 XP_005253955.1 11, 12 1179
          XM_006719445.2 XP_006719508.1 11, 12 1216
          XM_011538437.1 XP_011536739.1 11, 12 1234
          XM_011538439.1 XP_011536741.1 11, 12 1234
          XM_017019408.1 XP_016874897.1 12, 13 1499
          XM_017019409.1 XP_016874898.1 12, 13 1623
          XM_017019410.1 XP_016874899.1 12, 13 1619
          XM_017019411.1 XP_016874900.1 12, 13 1470
          XM_017019412.1 XP_016874901.1 11, 12 2310
          XM_017019413.1 XP_016874902.1 11, 12 2310
          GenBank mRNA AF135158.1 - 11, 12 1121
          AF179867.1 - 10, 11 1027
          AF181985.1 - 11, 12 1118
          AF263311.1 - 9, 10 826
          AK289558.1 - 11, 12 1261
          AL833460.1 - 7, 8 1413
          AL833731.1 - 3, 4 258
          BC002756.2 - 11, 12 1228 3630338
          BT007185.1 - 9, 10 826

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          DNA repair
          protein phosphorylation
          negative regulation of protein kinase activity
          cellular response to DNA damage stimulus
          mitotic G2 DNA damage checkpoint
          positive regulation of stress-activated MAPK cascade
          positive regulation of JUN kinase activity
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          protein autophosphorylation
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein kinase inhibitor activity
          protein binding
          ATP binding
          transferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0