Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNAS SIRNA Assays ›
          Gene Symbol:
          GNAS
          Gene Name:
          GNAS complex locus
          Gene Aliases:
          AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
          Chromosome Location:
          Chr.20: 58839681 - 58911196 on Build GRCh38
          UniGene ID:
          Hs.125898
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145435

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GNAS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000516.5 NP_000507.1 9, 10 1149
          NM_001077488.3 NP_001070956.1 10, 11 1152
          NM_001077489.3 NP_001070957.1 8, 9 1104
          NM_001077490.2 NP_001070958.1 9, 10 2939
          NM_001309840.1 NP_001296769.1 9, 10 815
          NM_001309861.1 NP_001296790.1 9, 10 860
          NM_016592.3 NP_057676.1 10, 11 1758
          NM_080425.3 NP_536350.2 9, 10 2939
          NM_080426.3 NP_536351.1 9, 10 1107
          XM_017027812.1 XP_016883301.1 10, 11 3209
          XM_017027813.1 XP_016883302.1 9, 10 3164
          XM_017027814.1 XP_016883303.1 8, 9 3161
          XM_017027815.1 XP_016883304.1 8, 9 632
          XM_017027816.1 XP_016883305.1 10, 11 1437
          XM_017027817.1 XP_016883306.1 8, 9 719
          XM_017027818.1 XP_016883307.1 8, 9 642
          XM_017027819.1 XP_016883308.1 8, 9 683
          XM_017027820.1 XP_016883309.1 8, 9 623
          GenBank mRNA AF105253.1 - 10, 11 1736
          AF493897.1 - 9, 10 725
          AF493898.1 - 9, 10 683
          AK026564.1 - 10, 11 919
          AK054862.1 - 7, 8 2013
          AK093534.1 - 8, 9 959
          AK122771.1 - 7, 8 1132
          AK225818.1 - 9, 10 848
          AK315860.1 - 9, 10 647
          AK315874.1 - 9, 10 828
          AL537118.3 - 9, 10 845
          BC002722.2 - 10, 11 843
          BC008855.2 - 11, 12 870
          BC022875.1 - 8, 9 571
          BC066923.1 - 8, 9 683
          BC089157.1 - 9, 10 999
          BC104928.1 - 9, 10 771
          BC108315.1 - 8, 9 945
          BT009905.1 - 9, 10 725
          KU178095.1 - 9, 10 725
          KU178096.1 - 9, 10 683
          M14631.1 - 10, 11 695
          X04408.1 - 9, 10 793
          X04409.1 - 9, 10 695
          X07036.1 - 9, 10 737
          X56009.1 - 9, 10 801

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          embryonic hindlimb morphogenesis
          multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          bone development
          hair follicle placode formation
          platelet aggregation
          genetic imprinting
          regulation of parathyroid hormone secretion
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pqldv:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0