Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDIT3 SIRNA Assays ›
          Gene Symbol:
          DDIT3
          Gene Name:
          DNA damage inducible transcript 3
          Gene Aliases:
          CEBPZ, CHOP, CHOP-10, CHOP10, GADD153
          Chromosome Location:
          Chr.12: 57516588 - 57520517 on Build GRCh38
          UniGene ID:
          Hs.505777
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146321

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDIT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195053.1 NP_001181982.1 4 817
          NM_001195054.1 NP_001181983.1 4 764
          NM_001195055.1 NP_001181984.1 4 741
          NM_001195056.1 NP_001181985.1 3 927
          NM_001195057.1 NP_001181986.1 4 646
          NM_004083.5 NP_004074.2 4 660
          GenBank mRNA AK316581.1 - 3 921
          AY461580.1 - 4 819
          BC003637.2 - 4 661 3530545
          BM464290.1 - 4 650
          BT006691.1 - 2 480
          S40706.1 - 4 651

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel maturation
          response to amphetamine
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          ER overload response
          response to unfolded protein
          cell cycle arrest
          aging
          response to nutrient
          Wnt signaling pathway
          positive regulation of interleukin-8 production
          negative regulation of CREB transcription factor activity
          response to endoplasmic reticulum stress
          PERK-mediated unfolded protein response
          ATF6-mediated unfolded protein response
          response to drug
          response to hydrogen peroxide
          response to starvation
          mRNA transcription from RNA polymerase II promoter
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neuron apoptotic process
          regulation of DNA-templated transcription in response to stress
          regulation of transcription involved in anterior/posterior axis specification
          cell redox homeostasis
          negative regulation of fat cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic organ development
          release of sequestered calcium ion into cytosol
          negative regulation of protein kinase B signaling
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of canonical Wnt signaling pathway
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          negative regulation of determination of dorsal identity
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          cAMP response element binding protein binding
          protein homodimerization activity
          leucine zipper domain binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0