Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146531
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
9, 10
1400
NM_004912.3
NP_004903.2
11, 12
2039
NM_194454.1
NP_919436.1
11, 12
1692
NM_194455.1
NP_919437.1
11, 12
2166
NM_194456.1
NP_919438.1
12, 13
1931
XM_005250660.3
XP_005250717.1
11, 12
1659
XM_005250662.3
XP_005250719.1
11, 12
1638
XM_005250665.3
XP_005250722.1
11, 12
1566
XM_005250666.3
XP_005250723.1
12, 13
1742
XM_005250667.2
XP_005250724.1
10, 11
1450
XM_005250668.3
XP_005250725.1
10, 11
1512
XM_005250669.3
XP_005250726.1
10, 11
1490
XM_006716161.3
XP_006716224.1
11, 12
1790
XM_006716162.3
XP_006716225.1
11, 12
1607
XM_006716163.3
XP_006716226.1
10, 11
1457
XM_011516651.2
XP_011514953.1
12, 13
1773
XM_011516653.2
XP_011514955.1
10, 11
1527
XM_011516654.2
XP_011514956.1
12, 13
1791
XM_011516655.2
XP_011514957.1
10, 11
1445
XM_011516656.2
XP_011514958.1
10, 11
1414
XM_011516657.2
XP_011514959.1
10, 11
1334
XM_011516658.2
XP_011514960.1
10, 11
1346
XM_011516659.2
XP_011514961.1
10, 11
1368
XM_011516660.2
XP_011514962.1
9, 10
1250
XM_011516661.2
XP_011514963.1
9, 10
1328
XM_017012755.1
XP_016868244.1
10, 11
1417
XM_017012756.1
XP_016868245.1
10, 11
1379
XM_017012757.1
XP_016868246.1
10, 11
1411
XM_017012758.1
XP_016868247.1
11, 12
1886
XM_017012759.1
XP_016868248.1
10, 11
1738
GenBank mRNA
AF296765.2
-
11, 12
1936
AF310133.1
-
11, 12
1631
AF388384.1
-
11, 12
2166
AK055305.1
-
9, 10
1400
AK056537.1
-
10, 11
1474
AK226131.1
-
4, 5
789
AY380057.1
-
11, 12
1631
AY380058.1
-
11, 12
1631
AY380059.1
-
11, 12
1631
AY380060.1
-
11, 12
1631
AY993944.1
-
8, 9
1147
BC094684.1
-
8, 9
1157
6174749
U90268.1
-
4, 5
551
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0