Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BRAF SIRNA Assays ›
          Gene Symbol:
          BRAF
          Gene Name:
          B-Raf proto-oncogene, serine/threonine kinase
          Gene Aliases:
          B-RAF1, B-raf, BRAF1, NS7, RAFB1
          Chromosome Location:
          Chr.7: 140719331 - 140924764 on Build GRCh38
          UniGene ID:
          Hs.550061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147343

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BRAF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004333.4 NP_004324.2 3 523
          XM_005250045.2 XP_005250102.1 3 470
          XM_017012558.1 XP_016868047.1 3 470
          XM_017012559.1 XP_016868048.1 3 470
          GenBank mRNA AK054801.1 - 1 1582
          AK314346.1 - 3 470
          BC101757.1 - 3 484 8069263
          BC112079.1 - 3 484 8327547
          M95712.2 - 3 523

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          myeloid progenitor cell differentiation
          protein phosphorylation
          visual learning
          organ morphogenesis
          positive regulation of gene expression
          negative regulation of fibroblast migration
          glucose transport
          thyroid gland development
          positive regulation of peptidyl-serine phosphorylation
          somatic stem cell population maintenance
          cellular response to drug
          regulation of cell proliferation
          negative regulation of apoptotic process
          CD4-positive, alpha-beta T cell differentiation
          positive T cell selection
          response to peptide hormone
          negative regulation of neuron apoptotic process
          thymus development
          positive regulation of axon regeneration
          positive regulation of axonogenesis
          protein heterooligomerization
          positive regulation of stress fiber assembly
          response to cAMP
          long-term synaptic potentiation
          head morphogenesis
          face development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          establishment of protein localization to membrane
          positive regulation of substrate adhesion-dependent cell spreading
          negative regulation of synaptic vesicle exocytosis
          negative regulation of endothelial cell apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          calcium ion binding
          protein binding
          ATP binding
          small GTPase binding
          mitogen-activated protein kinase kinase binding
          identical protein binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0