Search
Search

RTKN2| Gene Symbol: RTKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145307.3 | NP_660350.2 | 7 | 1002 | ||
| XM_011539456.2 | XP_011537758.1 | 7 | 1018 | |||
| XM_011539457.2 | XP_011537759.1 | 7 | 1018 | |||
| XM_011539460.2 | XP_011537762.1 | 7 | 1018 | |||
| XM_017015842.1 | XP_016871331.1 | 6 | 920 | |||
| XM_017015843.1 | XP_016871332.1 | 7 | 1018 | |||
| XM_017015844.1 | XP_016871333.1 | 7 | 1018 | |||
| GenBank mRNA | AK096671.1 | - | 2 | 117 | ||
| AY150309.1 | - | 7 | 846 | |||