Search
Search

OBFC1| Gene Symbol: OBFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024928.4 | NP_079204.2 | 8, 9 | 1125 | ||
| XM_006717976.3 | XP_006718039.1 | 8, 9 | 1125 | |||
| XM_011540184.2 | XP_011538486.1 | 8, 9 | 1125 | |||
| XM_017016669.1 | XP_016872158.1 | 7, 8 | 1292 | |||
| XM_017016670.1 | XP_016872159.1 | 8, 9 | 1125 | |||
| GenBank mRNA | AK026212.1 | - | 7, 8 | 950 | ||
| BC017400.2 | - | 8, 9 | 1046 | 3029792 | ||