Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKM SIRNA Assays ›
          Gene Symbol:
          Pkm C330027C09Rik
          Gene Name:
          pyruvate kinase, muscle
          RIKEN cDNA C330027C09 gene
          Gene Aliases:
          AA414905, AL024370, AL024424, Pk-2, Pk-3, Pk3, Pkm2
          AA408511, AU018569, Cip2a, Kiaa1524
          Chromosome Location:
          Chr.9: 59656368 - 59679375 on Build GRCm38
          Chromosome Location:
          Chr.16: 48994188 - 49019711 on Build GRCm38
          UniGene ID:
          Mm.326167
          Mm.24491
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 151118

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pkm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253883.1 NP_001240812.1 4 563
          NM_011099.3 NP_035229.2 4 563
          GenBank mRNA AK083076.1 - 4 356
          AK135397.1 - 4 356
          AK151724.1 - 4 429
          AK153483.1 - 4 429
          AK154695.1 - 4 356
          AK155110.1 - 4 356
          AK155655.1 - 4 359
          AK168943.1 - 4 356
          AK169872.1 - 4 335
          AK170892.1 - 4 359
          AK171023.1 - 4 356
          AK171033.1 - 4 359
          AK171106.1 - 4 360
          AK183643.1 - 3 142
          AK189659.1 - 3 142
          AK197420.1 - 142
          AK200951.1 - 4 337
          AK207130.1 - 3 142
          AK207766.1 - 3 142
          AK207924.1 - 3 142
          AK208026.1 - 3 142
          AK208591.1 - 3 142
          AK209065.1 - 3 142
          AK209125.1 - 3 142
          AK209411.1 - 3 142
          AK209972.1 - 3 142
          AK210823.1 - 3 142
          AK212924.1 - 3 142
          AK213742.1 - 3 142
          AK216406.1 - 3 142
          AK216783.1 - 3 142
          AK217024.1 - 163
          AK217172.1 - 3 142
          AK217300.1 - 3 142
          AK217516.1 - 3 142
          BC016619.1 - 4 323 3598842
          BC094663.1 - 4 302 6829532
          D38379.1 - 4 297
          X97047.1 - 3 274
          Gene Symbol: C330027C09Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC021874.1 - 16

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          glucose metabolic process
          glycolytic process
          ATP biosynthetic process
          metabolic process
          programmed cell death
          phosphorylation
          organ regeneration
          pyruvate biosynthetic process
          skeletal muscle tissue regeneration
          nucleotide binding
          magnesium ion binding
          catalytic activity
          pyruvate kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          MHC class II protein complex binding
          potassium ion binding
          ADP binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0