Search
Search

Irak4| Gene Symbol: Irak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029926.5 | NP_084202.2 | 4 | 577 | ||
| GenBank mRNA | AF445803.1 | - | 4 | 565 | ||
| AK020397.2 | - | 4 | 586 | |||
| AK028837.1 | - | 4 | 518 | |||
| AK029028.1 | - | 4 | 578 | |||
| AK139992.1 | - | 566 | ||||
| BC051676.1 | - | 4 | 570 | 30017484 | ||