Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LGALS3 SIRNA Assays ›
          Gene Symbol:
          Lgals3
          Gene Name:
          lectin, galactose binding, soluble 3
          Gene Aliases:
          GBP, L-34, Mac-2, gal3
          Chromosome Location:
          Chr.14: 47373860 - 47386167 on Build GRCm38
          UniGene ID:
          Mm.248615
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156106

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lgals3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145953.1 NP_001139425.1 6 790
          NM_010705.3 NP_034835.1 6 763
          GenBank mRNA AK008593.1 - 5 432
          AK089249.1 - 6 707
          AK149783.1 - 6 737
          AK155244.1 - 7 609
          AK155484.1 - 6 705
          AK159536.1 - 6 731
          AK170803.1 - 6 820
          BC132328.1 - 6 716 40130605
          BC138790.1 - 6 716 8861812
          BC145419.1 - 6 716 9053977
          J03723.1 - 4 663
          X16074.1 - 6 810
          X16834.1 - 6 709

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          immune system process
          monocyte chemotaxis
          mRNA processing
          signal transduction
          positive regulation of cell proliferation
          RNA splicing
          positive regulation of serotonin secretion
          cell differentiation
          extracellular matrix organization
          neutrophil chemotaxis
          regulation of T cell proliferation
          negative regulation of apoptotic process
          innate immune response
          positive regulation of angiogenesis
          negative regulation of endocytosis
          eosinophil chemotaxis
          macrophage chemotaxis
          negative regulation of T cell receptor signaling pathway
          positive chemotaxis
          regulation of T cell apoptotic process
          mononuclear cell migration
          positive regulation of mononuclear cell migration
          positive regulation of calcium ion import
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of cell proliferation in bone marrow
          negative regulation of immunological synapse formation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          negative regulation of extrinsic apoptotic signaling pathway
          protein binding
          IgE binding
          carbohydrate binding
          Fc-gamma receptor I complex binding
          chemoattractant activity
          laminin binding
          poly(A) RNA binding
          monosaccharide binding
          disaccharide binding
          advanced glycation end-product receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0