Search
Search

B3gnt3| Gene Symbol: B3gnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028189.3 | NP_082465.3 | 2 | 573 | ||
| GenBank mRNA | AK008674.1 | - | 612 | |||
| AK144117.1 | - | 563 | ||||
| AK144180.1 | - | 2 | 545 | |||
| AY037785.1 | - | 542 | ||||
| AY039028.1 | - | 1 | 492 | |||
| BC026418.1 | - | 2 | 565 | 4218953 | ||
| BY753510.1 | - | 2 | 573 | |||