Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026613.4 | NP_080889.3 | 6 | 1352 | ||
GenBank mRNA | AK011768.1 | - | 5 | 868 | ||
AK033555.1 | - | 6 | 1331 | |||
AK140947.1 | - | 3 | 3261 | |||
BC049229.1 | - | 6 | 1085 | |||
BC089458.1 | - | 8 | 1517 |