Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HTATIP2 SIRNA Assays ›
          Gene Symbol:
          Htatip2
          Gene Name:
          HIV-1 tat interactive protein 2, homolog (human)
          Gene Aliases:
          AW111545, Cc3, Tip30
          Chromosome Location:
          Chr.7: 49759045 - 49778364 on Build GRCm38
          UniGene ID:
          Mm.20801
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 184610

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Htatip2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146049.1 NP_001139521.1 6 924
          NM_001146050.1 NP_001139522.1 6 758
          NM_001146052.1 NP_001139524.1 5 748
          NM_001146053.1 NP_001139525.1 6 702
          NM_016865.3 NP_058561.2 6 868
          GenBank mRNA AF061972.1 - 5 852
          AK008630.1 - 5 743
          AK144837.1 - 6 902
          AK146400.1 - 5 563
          AK151254.1 - 6 789
          AK152418.1 - 6 789
          BC004083.1 - 6 821 3593077
          BC017372.1 - 5 714 4013057

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          apoptotic process
          multicellular organism development
          cell differentiation
          positive regulation of programmed cell death
          regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter during mitosis
          protein autophosphorylation
          nuclear import
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          protein serine/threonine kinase activity
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          3-ketoglucose-reductase activity
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0