Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009441.2 | NP_033467.2 | 20 | 1822 | ||
GenBank mRNA | AB008516.1 | - | 19 | 1699 | ||
AK053765.1 | - | 20 | 1926 | |||
AK084201.1 | - | 10 | 857 | |||
AK088273.1 | - | 19 | 1769 | |||
AK165850.1 | - | 9 | 730 | |||
AK166120.1 | - | 10 | 762 | |||
AK166164.1 | - | 11 | 911 | |||
AK220495.1 | - | 20 | 1864 | |||
BC019173.1 | - | 20 | 1791 | |||
BC037654.1 | - | 19 | 1724 | |||
BC057207.1 | - | 19 | 1726 | |||
BC145489.1 | - | 15 | 1342 |