Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TOP2A SIRNA Assays ›
Gene Symbol:
Top2a
Gene Name:
topoisomerase (DNA) II alpha
Gene Aliases:
Top-2
Chromosome Location:
Chr.11: 98992947 - 99024206 on Build GRCm38
UniGene ID:
Mm.4237
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
187389
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Top2a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011623.2
NP_035753.2
8
1055
GenBank mRNA
BC008170.1
-
8
1043
BC057873.1
-
8
1025
D12513.1
-
8
981
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_022183, Z19552.
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
resolution of meiotic recombination intermediates
sister chromatid segregation
hematopoietic progenitor cell differentiation
DNA metabolic process
DNA topological change
DNA ligation
DNA unwinding involved in DNA replication
cellular response to DNA damage stimulus
chromosome segregation
chromosome condensation
apoptotic chromosome condensation
embryonic cleavage
regulation of circadian rhythm
positive regulation of apoptotic process
mitotic DNA integrity checkpoint
positive regulation of viral genome replication
positive regulation of single stranded viral RNA replication via double stranded DNA intermediate
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
nucleotide binding
magnesium ion binding
DNA binding
chromatin binding
DNA topoisomerase activity
DNA topoisomerase type II (ATP-hydrolyzing) activity
protein kinase C binding
ATP binding
protein C-terminus binding
DNA-dependent ATPase activity
drug binding
DNA binding, bending
isomerase activity
enzyme binding
protein homodimerization activity
histone deacetylase binding
ubiquitin binding
sequence-specific DNA binding
structure-specific DNA binding
poly(A) RNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0