Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017007.1 | NP_058703.1 | 2, 3 | 257 | ||
GenBank mRNA | GQ468528.1 | - | 1, 2 | 436 | ||
M34445.1 | - | 2, 3 | 257 | |||
M38350.1 | - | 2, 3 | 174 | |||
M76177.1 | - | 2, 3 | 258 | |||
X57572.1 | - | 2, 3 | 201 | |||
X57573.1 | - | 2, 3 | 257 |