Search
Search

LVRN| Gene Symbol: LVRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173800.4 | NP_776161.3 | 12 | 2104 | ||
| XM_011543263.2 | XP_011541565.1 | 12 | 2118 | |||
| XM_011543264.2 | XP_011541566.1 | 12 | 2118 | |||
| GenBank mRNA | AK075131.1 | - | 7 | 639 | ||
| AK291655.1 | - | 12 | 2118 | |||
| AY560010.1 | - | 12 | 1992 | |||
| BC036440.1 | - | 7 | 757 | |||
| BC045809.1 | - | 7 | 732 | |||
| BC060869.1 | - | 13 | 2031 | 30346049 | ||
| BC068560.1 | - | 12 | 2104 | 30341970 | ||
| BC070028.1 | - | 7 | 752 | |||
| BC094716.1 | - | 7 | 752 | |||
| BC109022.1 | - | 12 | 1968 | 40008501 | ||
| BC109023.1 | - | 12 | 1968 | 40008502 | ||