Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2I SIRNA Assays ›
          Gene Symbol:
          UBE2I
          Gene Name:
          ubiquitin conjugating enzyme E2 I
          Gene Aliases:
          C358B7.1, P18, UBC9
          Chromosome Location:
          Chr.16: 1309153 - 1327018 on Build GRCh38
          UniGene ID:
          Hs.302903
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214058

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE2I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003345.4 NP_003336.1 6 562
          NM_194259.2 NP_919235.1 7 819
          NM_194260.2 NP_919236.1 6 682
          NM_194261.2 NP_919237.1 6 534
          XM_017023640.1 XP_016879129.1 6 959
          GenBank mRNA AB208988.1 - 6 643
          AJ002385.1 - 6 482
          AK291118.1 - 6 553
          BC000427.2 - 6 466 2819764
          BC004429.1 - 6 466 2819764
          BC051289.1 - 6 496
          BQ230918.1 - 1 52
          BT006932.1 - 5 404
          BU146357.1 - 2 135
          BU543455.1 - 6 554
          CD249028.1 - 6 503
          CX788607.1 - 6 638
          D45050.1 - 6 521
          U29092.1 - 5 428
          U31882.1 - 6 497
          U31933.1 - 6 522
          U38785.1 - 6 496
          U45328.1 - 6 527
          U66818.1 - 6 491
          U66867.1 - 6 1210
          X96427.1 - 6 527

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cellular protein modification process
          ubiquitin-dependent protein catabolic process
          chromosome segregation
          mitotic nuclear division
          viral process
          protein sumoylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of transcription, DNA-templated
          cell division
          global genome nucleotide-excision repair
          positive regulation of SUMO transferase activity
          protein binding
          ATP binding
          transcription factor binding
          ligase activity
          SUMO transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          HLH domain binding
          poly(A) RNA binding
          ubiquitin protein ligase activity
          SUMO conjugating enzyme activity
          RING-like zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zkdp8:80/100.66.75.244:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0