Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC78 SIRNA Assays ›
Gene Symbol:
CCDC78
Gene Name:
coiled-coil domain containing 78
Gene Aliases:
C16orf25, CNM4, JFP10, hsCCDC78
Chromosome Location:
Chr.16: 722578 - 726880 on Build GRCh38
UniGene ID:
Hs.381943
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215901
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC78
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031737.2
NP_001026907.2
13, 14
1411
XM_006720838.1
XP_006720901.1
11, 12
1692
XM_006720843.3
XP_006720906.1
12, 13
1849
XM_011522356.1
XP_011520658.1
8, 9
1917
XM_011522357.1
XP_011520659.1
10, 11
1905
XM_011522359.1
XP_011520661.1
8, 9
1884
XM_011522360.1
XP_011520662.1
10, 11
1872
XM_011522361.1
XP_011520663.1
9, 10
1845
XM_011522362.1
XP_011520664.1
9
1973
XM_011522363.1
XP_011520665.1
9, 10
1841
XM_011522365.1
XP_011520667.1
9, 10
1704
XM_011522366.1
XP_011520668.1
9, 10
1682
XM_011522367.1
XP_011520669.1
8, 9
1536
XM_011522368.1
XP_011520670.1
10, 11
1524
XM_011522369.1
XP_011520671.1
10, 11
1482
XM_011522370.1
XP_011520672.1
9, 10
1301
XM_011522371.2
XP_011520673.1
9, 10
1293
XM_017022930.1
XP_016878419.1
12, 13
1587
XM_017022931.1
XP_016878420.1
2, 3
1098
XM_017022932.1
XP_016878421.1
5, 6
939
GenBank mRNA
AK057178.1
-
12, 13
1653
AK091831.1
-
9, 10
1878
AK127495.1
-
4
2855
AK128538.1
-
4
3260
AK128575.1
-
4, 5
3054
BC042110.1
-
13, 14
1397
5177109
Back To Top
More Information
Gene Ontology Categories:
Function(s)
skeletal muscle contraction
cell projection organization
de novo centriole assembly
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lfs6t:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0