Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF25 SIRNA Assays ›
Gene Symbol:
TCF25
Gene Name:
transcription factor 25
Gene Aliases:
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049
Chromosome Location:
Chr.16: 89873583 - 89911384 on Build GRCh38
UniGene ID:
Hs.415342
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22557
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014972.2
NP_055787.1
12
1428
XM_005256297.2
XP_005256354.1
12
1422
XM_005256298.2
XP_005256355.1
12
1422
XM_005256299.2
XP_005256356.1
12
1377
XM_011522956.2
XP_011521258.1
12
1437
XM_011522957.2
XP_011521259.1
12
1437
XM_011522958.2
XP_011521260.1
12
1392
XM_011522959.2
XP_011521261.1
11
1371
XM_011522960.2
XP_011521262.1
12
1437
XM_011522961.2
XP_011521263.1
13
1606
XM_017023047.1
XP_016878536.1
11
1356
XM_017023048.1
XP_016878537.1
12
1377
XM_017023049.1
XP_016878538.1
12
1392
XM_017023050.1
XP_016878539.1
12
1377
XM_017023051.1
XP_016878540.1
11
1356
XM_017023052.1
XP_016878541.1
11
1311
XM_017023053.1
XP_016878542.1
13
2044
XM_017023054.1
XP_016878543.1
13
1591
XM_017023055.1
XP_016878544.1
13
1546
XM_017023056.1
XP_016878545.1
13
1598
XM_017023057.1
XP_016878546.1
13
2029
XM_017023058.1
XP_016878547.1
12
1963
GenBank mRNA
AB028972.1
-
10
969
AF130092.1
-
9
930
AF322111.1
-
12
1442
BC000959.1
-
12
1351
3445789
BC009349.2
-
12
1373
4136178
BC011884.2
-
12
1402
3678670
DQ321703.1
-
12
1442
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
heart development
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0