Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 10 | 4752 | ||
NM_015264.1 | NP_056079.1 | 10 | 4824 | |||
XM_011530017.2 | XP_011528319.1 | 10 | 4787 | |||
XM_017028672.1 | XP_016884161.1 | 10 | 4796 |