Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF4G3 SIRNA Assays ›
          Gene Symbol:
          EIF4G3
          Gene Name:
          eukaryotic translation initiation factor 4 gamma 3
          Gene Aliases:
          eIF-4G 3, eIF4G 3, eIF4GII
          Chromosome Location:
          Chr.1: 20806292 - 21177320 on Build GRCh38
          UniGene ID:
          Hs.467084
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 266747

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF4G3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198801.1 NP_001185730.1 1, 2 159
          NM_001198802.1 NP_001185731.1 1, 2 159
          XM_011542356.2 XP_011540658.1 1, 2 591
          XM_011542361.2 XP_011540663.1 1, 2 591
          XM_011542370.1 XP_011540672.1 1, 2 591
          XM_011542375.2 XP_011540677.1 1, 2 591
          XM_017002672.1 XP_016858161.1 1, 2 591
          XM_017002673.1 XP_016858162.1 1, 2 591
          XM_017002674.1 XP_016858163.1 1, 2 591
          XM_017002675.1 XP_016858164.1 1, 2 591
          XM_017002676.1 XP_016858165.1 1, 2 591
          XM_017002678.1 XP_016858167.1 1, 2 591
          XM_017002679.1 XP_016858168.1 1, 2 591
          XM_017002681.1 XP_016858170.1 1, 2 591
          XM_017002682.1 XP_016858171.1 1, 2 591
          XM_017002683.1 XP_016858172.1 1, 2 591
          XM_017002684.1 XP_016858173.1 1, 2 591
          XM_017002685.1 XP_016858174.1 1, 2 591
          XM_017002686.1 XP_016858175.1 1, 2 591
          XM_017002687.1 XP_016858176.1 1, 2 591
          XM_017002689.1 XP_016858178.1 1, 2 591
          XM_017002690.1 XP_016858179.1 1, 2 591
          XM_017002691.1 XP_016858180.1 1, 2 591
          XM_017002692.1 XP_016858181.1 1, 2 591
          XM_017002693.1 XP_016858182.1 1, 2 591
          XM_017002694.1 XP_016858183.1 1, 2 591
          XM_017002695.1 XP_016858184.1 1, 2 591
          XM_017002696.1 XP_016858185.1 1, 2 591
          XM_017002697.1 XP_016858186.1 1, 2 591
          XM_017002698.1 XP_016858187.1 1, 2 591
          XM_017002699.1 XP_016858188.1 1, 2 591
          XM_017002700.1 XP_016858189.1 1, 2 591
          XM_017002701.1 XP_016858190.1 1, 2 591
          XM_017002702.1 XP_016858191.1 1, 2 591
          XM_017002704.1 XP_016858193.1 1, 2 591
          XM_017002705.1 XP_016858194.1 1, 2 591
          XM_017002706.1 XP_016858195.1 1, 2 591
          XM_017002707.1 XP_016858196.1 1, 2 591
          XM_017002708.1 XP_016858197.1 1, 2 591
          XM_017002709.1 XP_016858198.1 1, 2 591
          XM_017002710.1 XP_016858199.1 1, 2 591
          XM_017002711.1 XP_016858200.1 1, 2 591
          XM_017002712.1 XP_016858201.1 1, 2 591
          GenBank mRNA AB209119.1 - 1, 2 121
          CN402454.1 - 1, 2 160

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001256198, NM_001256195, NM_172703, BC072600, BC082549, and others.

          Panther Classification:

          Molecular Function -

          translation factor translation initiation factor translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational initiation
          regulation of translational initiation
          viral process
          RNA cap binding
          translation initiation factor activity
          translation factor activity, RNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0