Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTN SIRNA Assays ›
Gene Symbol:
Ttn
Gene Name:
titin
Gene Aliases:
1100001C23Rik, 2310036G12Rik, 2310057K23Rik, 2310074I15Rik, AF006999, AV006427, D330041I19Rik, D830007G01Rik, L56, mdm, shru
Chromosome Location:
Chr.2: 76703980 - 76982557 on Build GRCm38
UniGene ID:
Mm.373672
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
282819
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ttn
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011652.3
NP_035782.3
22
3818
NM_028004.2
NP_082280.2
21
3680
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
heart morphogenesis
cardiac muscle hypertrophy
muscle contraction
adult heart development
peptidyl-tyrosine phosphorylation
ventricular system development
skeletal muscle thin filament assembly
skeletal muscle myosin thick filament assembly
detection of muscle stretch
forward locomotion
sarcomere organization
regulation of protein kinase activity
cardiac muscle fiber development
sarcomerogenesis
regulation of catalytic activity
response to calcium ion
striated muscle cell development
cardiac myofibril assembly
cardiac muscle tissue morphogenesis
cardiac muscle contraction
heart growth
protease binding
protein serine/threonine kinase activity
protein tyrosine kinase activity
structural constituent of cytoskeleton
calcium ion binding
protein binding
calmodulin binding
structural constituent of muscle
enzyme binding
protein kinase binding
telethonin binding
identical protein binding
actinin binding
protein self-association
actin filament binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-rpnmj:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0