Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARFRP1 SIRNA Assays ›
Gene Symbol:
ARFRP1
Gene Name:
ADP ribosylation factor related protein 1
Gene Aliases:
ARL18, ARP, Arp1
Chromosome Location:
Chr.20: 63698642 - 63708013 on Build GRCh38
UniGene ID:
Hs.389277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289415
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARFRP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134758.3
NP_001128230.1
4, 5
405
NM_001267544.2
NP_001254473.1
4, 5
410
NM_001267545.2
NP_001254474.1
4, 5
405
NM_001267546.2
NP_001254475.1
3, 4
311
NM_001267547.2
NP_001254476.1
4, 5
410
NM_001267548.2
NP_001254477.1
4, 5
307
NM_001267549.2
NP_001254478.1
4, 5
410
NM_003224.5
NP_003215.1
4, 5
405
NR_051954.2
4, 5
405
NR_051955.2
4, 5
410
NR_051956.2
4, 5
410
NR_051957.2
4, 5
405
NR_051958.2
4, 5
405
XM_011528482.2
XP_011526784.1
4, 5
342
XM_011528483.1
XP_011526785.1
4, 5
472
XM_017027576.1
XP_016883065.1
4, 5
286
GenBank mRNA
AK095962.1
-
3, 4
824
AL522492.3
-
3, 4
262
AL533893.3
-
4, 5
395
BC021023.1
-
4, 5
290
BC092480.1
-
4, 5
291
30531333
BC093967.1
-
4, 5
308
7939812
BC143447.1
-
4, 5
308
9051955
BC143448.1
-
4, 5
313
9051956
BC143467.1
-
4, 5
313
9051975
BC143477.1
-
4, 5
313
BI524032.1
-
4, 5
404
BI819763.1
-
4, 5
279
BM546222.1
-
330
BM549361.1
-
373
BT019624.1
-
3, 4
257
BT019663.1
-
3, 4
257
BU526941.1
-
4, 5
347
BX342690.2
-
4, 5
291
BX342805.2
-
4, 5
384
BX463457.2
-
4, 5
307
CA489361.1
-
3, 4
249
CR456752.1
-
3, 4
257
X91504.1
-
3, 4
268
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
small GTPase mediated signal transduction
gastrulation
protein localization to Golgi apparatus
retrograde transport, endosome to Golgi
Golgi to plasma membrane protein transport
GTPase activity
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4hhk6:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0