Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARMC9 SIRNA Assays ›
Gene Symbol:
ARMC9
Gene Name:
armadillo repeat containing 9
Gene Aliases:
ARM, KU-MEL-1, NS21
Chromosome Location:
Chr.2: 231198546 - 231394991 on Build GRCh38
UniGene ID:
Hs.162411
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32383
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARMC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271466.2
NP_001258395.1
2, 3
227
NM_001291656.1
NP_001278585.1
3, 4
380
NM_025139.5
NP_079415.3
3, 4
382
XM_011511905.1
XP_011510207.1
3, 4
300
XM_011511906.1
XP_011510208.1
3, 4
300
XM_011511907.1
XP_011510209.1
3, 4
300
XM_011511908.2
XP_011510210.1
3, 4
300
XM_011511909.2
XP_011510211.1
3, 4
300
XM_011511910.1
XP_011510212.1
3, 4
300
XM_011511911.1
XP_011510213.1
3, 4
300
XM_011511912.1
XP_011510214.1
3, 4
300
XM_011511913.2
XP_011510215.1
3, 4
300
XM_011511914.2
XP_011510216.1
3, 4
300
XM_011511915.2
XP_011510217.1
3, 4
300
XM_011511916.2
XP_011510218.1
3, 4
300
XM_017005018.1
XP_016860507.1
3, 4
558
XM_017005019.1
XP_016860508.1
3, 4
476
XM_017005020.1
XP_016860509.1
3, 4
558
XM_017005021.1
XP_016860510.1
3, 4
300
XM_017005022.1
XP_016860511.1
3, 4
300
XM_017005023.1
XP_016860512.1
3, 4
558
XM_017005024.1
XP_016860513.1
3, 4
300
XM_017005025.1
XP_016860514.1
3, 4
300
XM_017005026.1
XP_016860515.1
3, 4
499
GenBank mRNA
AK022646.1
-
1, 2
84
AK309395.1
-
289
AY219922.1
-
3, 4
261
AY929062.1
-
3, 4
254
BC065271.1
-
2, 3
214
6165433
BM563736.1
-
390
BX537956.1
-
3, 4
298
DA030137.1
-
3, 4
301
HY022484.1
-
3, 4
348
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0