Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159352.1 | NP_001152824.1 | 7 | 1386 | ||
NM_032044.3 | NP_114433.1 | 6 | 1212 | |||
GenBank mRNA | AF254415.1 | - | 6 | 1127 | ||
AF345934.1 | - | 6 | 1082 | |||
AY007243.1 | - | 6 | 1068 | |||
AY126670.1 | - | 7 | 1386 | |||
AY126671.1 | - | 5 | 620 | |||
BC017089.2 | - | 6 | 1084 | 3850358 |