Search
Search

KMT5C| Gene Symbol: KMT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032701.3 | NP_116090.2 | 8 | 968 | ||
| XM_005259338.3 | XP_005259395.1 | 7 | 1293 | |||
| XM_006723442.3 | XP_006723505.1 | 6 | 701 | |||
| XM_011527415.2 | XP_011525717.1 | 8 | 1488 | |||
| XM_017027388.1 | XP_016882877.1 | 6 | 1749 | |||
| GenBank mRNA | BC005842.2 | - | 6 | 702 | 2821053 | |
| BC019313.2 | - | 6 | 697 | 2821053 | ||
| BC044889.1 | - | 8 | 925 | 3461265 | ||