Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082484.2 | NP_001075953.1 | 12 | 1919 | ||
NM_029721.2 | NP_083997.1 | 12 | 2713 | |||
GenBank mRNA | AK041201.1 | - | 5 | 1416 | ||
AK043074.1 | - | 3 | 303 | |||
AK122296.1 | - | 12 | 2524 | |||
AK147452.1 | - | 12 | 2699 | |||
AK164500.1 | - | 1 | 147 |