Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRG1 SIRNA Assays ›
          Gene Symbol:
          Nrg1
          Gene Name:
          neuregulin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 62969988 - 64057434 on Build Rnor_6.0
          UniGene ID:
          Rn.37438
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 53760

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nrg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271123.1 NP_001258052.1 5 836
          NM_001271124.1 NP_001258053.1 5 836
          NM_001271125.1 NP_001258054.1 5 836
          NM_001271126.1 NP_001258055.1 5 836
          NM_001271127.1 NP_001258056.1 5 836
          NM_001271128.1 NP_001258057.1 5 836
          NM_001271129.1 NP_001258058.1 5 836
          NM_001271130.1 NP_001258059.1 5 906
          NM_031588.2 NP_113776.2 5 836
          GenBank mRNA AF194993.1 - 5 906
          AF194995.1 - 5 739
          AF194996.1 - 5 739
          AF194997.1 - 5 739
          AY973241.1 - 3 228
          AY973243.1 - 3 228
          AY973244.1 - 3 192
          AY995221.1 - 5 492
          M92430.1 - 5 536
          U02315.1 - 5 461
          U02316.1 - 5 836
          U02317.1 - 5 836
          U02318.1 - 5 836
          U02319.1 - 5 836
          U02320.1 - 5 836
          U02321.1 - 5 836
          U02322.1 - 5 836
          U02323.1 - 5 836
          U02324.1 - 5 536

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cell morphogenesis
          cardiac conduction system development
          ventricular trabecula myocardium morphogenesis
          transmembrane receptor protein tyrosine kinase signaling pathway
          activation of transmembrane receptor protein tyrosine kinase activity
          nervous system development
          synapse assembly
          peripheral nervous system development
          heart development
          muscle organ development
          locomotory behavior
          regulation of blood pressure
          cell proliferation
          glial cell differentiation
          negative regulation of cardiac muscle cell apoptotic process
          glucose transport
          cell migration
          glial cell fate commitment
          cell differentiation
          positive regulation of cell growth
          intracellular signal transduction
          ERBB signaling pathway
          wound healing
          negative regulation of protein catabolic process
          myelination
          regulation of protein heterodimerization activity
          cellular protein complex disassembly
          neurotransmitter receptor metabolic process
          regulation of cell differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of Ras protein signal transduction
          animal organ development
          neuron fate commitment
          negative regulation of secretion
          positive regulation of protein kinase B signaling
          ventricular cardiac muscle cell differentiation
          positive regulation of cardiac muscle cell proliferation
          cardiac muscle cell myoblast differentiation
          endocardial cell differentiation
          positive regulation of protein tyrosine kinase activity
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of cardiac muscle cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          transcription cofactor activity
          receptor binding
          ErbB-2 class receptor binding
          growth factor activity
          protein tyrosine kinase activator activity
          ErbB-3 class receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0